Anti-F10 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB100441-100 ORB100441-200
BIRBORB100441-100EA 0 EUR
BIRBORB100441-100 BIRBORB100441-200
Anti-F10 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: F10 (coagulation factor X)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR