Anti-IgG Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB100479-100EA 0 EUR
BIRBORB100479-100
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR