Anti-HCFC1 Rabbit polyclonal antibody

Supplier: Biorbyt
BIRBORB100636-200EA 0 EUR
BIRBORB100636-200 BIRBORB100636-100
Anti-HCFC1 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: HCFC1 (host cell factor C1 (VP16-accessory protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR