Anti-SEMA6D Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100716-100 ORB100716-200
BIRBORB100716-100EA 0 EUR
BIRBORB100716-100 BIRBORB100716-200
Anti-SEMA6D Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SEMA6D (sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR