Anti-RNASE4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100741-100 ORB100741-200
BIRBORB100741-100EA 0 EUR
BIRBORB100741-100 BIRBORB100741-200
Anti-RNASE4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: RNASE4 (ribonuclease, RNase A family, 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR