Anti-SOAT1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100781-100 ORB100781-20 ORB100781-200 ORB100781-5
BIRBORB100781-100EA 0 EUR
BIRBORB100781-100 BIRBORB100781-20 BIRBORB100781-200 BIRBORB100781-5
Anti-SOAT1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SOAT1 (sterol O-acyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR