Anti-BMP5 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB100930-100 ORB100930-200
BIRBORB100930-100EA 0 EUR
BIRBORB100930-100 BIRBORB100930-200
Anti-BMP5 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: BMP5 (bone morphogenetic protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR