Anti-C3AR1 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB101135-100 ORB101135-200
BIRBORB101135-100EA 0 EUR
BIRBORB101135-100 BIRBORB101135-200
Anti-C3AR1 Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: C3AR1 (complement component 3a receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR