Anti-SLC25A6 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101369-100 ORB101369-200
BIRBORB101369-100EA 0 EUR
BIRBORB101369-100 BIRBORB101369-200
Anti-SLC25A6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SLC25A6 (solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR