Anti-RSRC2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101482-100 ORB101482-200
BIRBORB101482-100EA 0 EUR
BIRBORB101482-100 BIRBORB101482-200
Anti-RSRC2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; IC 1:100-500; Immunofluorescence: 1:50-200

Type: Primary
Antigen: RSRC2 (arginine/serine-rich coiled-coil 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR