Anti-P4HTM Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101845-100 ORB101845-200
BIRBORB101845-100EA 0 EUR
BIRBORB101845-100 BIRBORB101845-200
Anti-P4HTM Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: P4HTM (prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR