Anti-Sortilin Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB101927-100 ORB101927-200
BIRBORB101927-100EA 0 EUR
BIRBORB101927-100 BIRBORB101927-200
Anti-Sortilin Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: Sortilin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR