Anti-IgM Mouse Polyclonal Antibody

Supplier: Biorbyt
ORB101945-100 ORB101945-200
BIRBORB101945-100EA 0 EUR
BIRBORB101945-100 BIRBORB101945-200
Anti-IgM Mouse Polyclonal Antibody
Antibodies
Western Blot: 1:1000-5000; ELISA: 1:2000-10000; Immunochemistry: 1:200-1000; IC 1:200-1000

Type: Secondary
Antigen: IgM (Human) (human IgM)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR