Anti-SEPT10 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB101988-100
BIRBORB101988-100EA 0 EUR
BIRBORB101988-100
Anti-SEPT10 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: SEPT10 (septin 10)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR