Anti-AIFM3 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
ORB102049-100
BIRBORB102049-100EA 0 EUR
BIRBORB102049-100
Anti-AIFM3 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: AIFM3 (apoptosis-inducing factor, mitochondrion-associated, 3)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR