Anti-GGCX Goat polyclonal antibody unconjugated

Supplier: Abcam
ABCAAB106924-100EA 833 EUR
ABCAAB106924-100
Anti-GGCX Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: GGCX (gamma-glutamyl carboxylase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR