Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 405)

Supplier: Novus Biologicals
NOVUNB110-40765VEA 656 EUR
NOVUNB110-40765V
Anti-PLIN3 Rabbit Polyclonal Antibody (DyLight® 405)
Antibodies
Type: Primary
Antigen: PLIN3 (perilipin 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR