Anti-STRAP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NOVUNBP2-24740EA 425 EUR
NOVUNBP2-24740
Anti-STRAP Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STRAP (serine/threonine kinase receptor associated protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR