Anti-EhpB6 Mouse Monoclonal Antibody [clone: 2A6B9]

Supplier: Thermo Fisher Scientific
PIEAMA515384EA 0 EUR
PIEAMA515384
Anti-EhpB6 Mouse Monoclonal Antibody [clone: 2A6B9]
Antibodies
MA5-15384 targets EhpB6 in IHC and WB applications and shows reactivity with Human samples. The MA5-15384 immunogen is purified recombinant fragment of EphB6 (aa601-750) expressed in E. Coli.<br>. Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase family. The ephrin receptor encoded by this gene lacks the kinase activity of most receptor tyrosine kinases and binds to ephrin-B ligands.

Type: Primary
Antigen: EhpB6
Clonality: Monoclonal
Clone: 2A6B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR