Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
Global Web is currently experiencing issues, technical teams are working to correct the problem.

Anti-EhpB1 Mouse Monoclonal Antibody [clone: 5F10A4]

Supplier: Thermo Scientific
MA5-15385 targets EhpB1 in IHC and WB applications and shows reactivity with Human samples. The MA5-15385 immunogen is purified recombinant fragment of EphB1 (aa19-133) expressed in E. Coli. <br>. MA5-15385 detects EhpB1 which has a predicted molecular weight of approximately 110kDa. Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase family. The protein encoded by this gene is a receptor for ephrin-B family members.

Type: Primary
Antigen: EhpB1
Clonality: Monoclonal
Clone: 5F10A4
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG1
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAMA515385 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol EhpB1
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 5F10A4
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms ELK, EPHB1, EPHT2, Hek6, FLJ37986, NET
Gene ID 2047
Format ascites
Immunogen Purified recombinant fragment of EphB1 (aa19-133) expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty