Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CANX Mouse Monoclonal Antibody [clone: 3H4A7]

Supplier: Thermo Scientific
MA5-15389 targets Calnexin in IF and WB applications and shows reactivity with Human samples. The MA5-15389 immunogen is synthetic peptide corresponding to aa (CEAAEERPWLWVVYILTVAL) of human Calnexin, conjugated to KLH. MA5-15389 detects Calnexin which has a predicted molecular weight of approximately 90kDa. This gene encodes a member of the calnexin family of molecular chaperones. The encoded protein is a calcium-binding, endoplasmic reticulum -associated protein that interacts transiently with newly synthesized N-linked glycoproteins, facilitating protein folding and assembly. It may also play a central role in the quality control of protein folding by retaining incorrectly folded protein subunits within the ER for degradation. Alternatively spliced transcript variants encoding the same protein have been described.

Type: Primary
Antigen: CANX
Clonality: Monoclonal
Clone: 3H4A7
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAMA515389 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol CANX
Antigen name Calnexin
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 3H4A7
Host Mouse
Isotype IgG2b
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms CNX, P90, CANX, IP90, FLJ26570
Gene ID 821
Format ascites
Immunogen Synthetic peptide corresponding to aa (CEAAEERPWLWVVYILTVAL) of human Calnexin, conjugated to KLH.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty