Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EPHB3 Mouse Monoclonal Antibody [clone: 4A122D1]

Supplier: Thermo Scientific
MA5-15394 targets EphB3 in IHC and WB applications and shows reactivity with Human samples. The MA5-15394 immunogen is purified recombinant fragment of EphB3 (aa39-212) expressed in E. Coli. MA5-15394 has been successfully used in immunohistochemistry analysis of Ephb3 in mouse E16.5 frozen lung sections. Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B class, which are transmembrane proteins. The Eph family of receptors are divided into two groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase family. This gene encodes a receptor for ephrin-B family members.

Type: Primary
Antigen: EPHB3
Clonality: Monoclonal
Clone: 4A122D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAMA515394 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol EPHB3
Antigen name EPH Receptor B3
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 4A122D1
Host Mouse
Isotype IgG2a
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms EPHB3, ETK2, TYRO6, HEK2
Gene ID 2049
Format ascites
Immunogen Purified recombinant fragment of EphB3 (aa39-212) expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty