Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FBLN5 Mouse Monoclonal Antibody [clone: 1G6A4]

Supplier: Thermo Scientific
MA5-15395 targets FBLN5 in IHC and WB applications and shows reactivity with Human samples. The MA5-15395 immunogen is purified recombinant fragment of FBLN5 (aa242-448) expressed in E. Coli.<br>. The protein encoded by this gene is a secreted, extracellular matrix protein containing an Arg-Gly-Asp motif and calcium-binding EGF-like domains. It promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. It is prominently expressed in developing arteries but less so in adult vessels. However, its expression is reinduced in balloon-injured vessels and atherosclerotic lesions, notably in intimal vascular smooth muscle cells and endothelial cells. Therefore, the protein encoded by this gene may play a role in vascular development and remodeling. Defects in this gene are a cause of autosomal dominant cutis laxa, autosomal recessive cutis laxa type I , and age-related macular degeneration type 3 .

Type: Primary
Antigen: FBLN5
Clonality: Monoclonal
Clone: 1G6A4
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG1
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAMA515395 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol FBLN5
Antigen name Fibulin 5
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1G6A4
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms EVEC, FLJ90059, ARMD3, DANCE, FIBL-5, UP50, FBLN5
Gene ID 10516
Format ascites
Immunogen Purified recombinant fragment of FBLN5 (aa242-448) expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty