Anti-SRC Mouse Monoclonal Antibody [clone: 4F1E8]

Supplier: Thermo Fisher Scientific
PIEAMA515398EA 0 EUR
PIEAMA515398
Anti-SRC Mouse Monoclonal Antibody [clone: 4F1E8]
Antibodies
MA5-15398 targets SRC in WB applications and shows reactivity with Human samples. The MA5-15398 immunogen is purified recombinant fragment of SRC (aa10-193) expressed in E. Coli. This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene.

Type: Primary
Antigen: SRC
Clonality: Monoclonal
Clone: 4F1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR