Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IGFBP2 Mouse Monoclonal Antibody [clone: 1F6F6]

Supplier: Thermo Scientific
MA5-15400 targets IGFBP2 in IHC and WB applications and shows reactivity with Human samples. The MA5-15400 immunogen is purified recombinant fragment of protein IGFBP2 (aa180-328) expressed in E. Coli. <br>. IGFBP2: insulin-like growth factor binding protein 2. IGFBP2 is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of other cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.

Type: Primary
Antigen: IGFBP2
Clonality: Monoclonal
Clone: 1F6F6
Conjugation: Unconjugated
Host: Mouse
Isotype: IgG1
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAMA515400 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol IGFBP2
Antigen name Insulin-Like Growth Factor Binding Protein 2
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 1F6F6
Host Mouse
Isotype IgG1
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms IBP2, IGF-BP53, IGFBP2
Gene ID 3485
Format ascites
Immunogen Purified recombinant fragment of protein IGFBP2 (aa180-328) expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty