Anti-TYRO3 Mouse Monoclonal Antibody [clone: 6D6F10]

Supplier: Thermo Fisher Scientific
PIEAMA515404EA 0 EUR
PIEAMA515404
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 6D6F10]
Antibodies
MA5-15404 targets Tyro3 in IF applications and shows reactivity with Human samples. The MA5-15404 immunogen is purified recombinant fragment of Tyro3 (aa138-321) expressed in E. Coli. The gene is part of a 3-member transmembrane receptor kinase receptor family with a processed pseudogene distal on chromosome 15. The encoded protein is activated by the products of the growth arrest-specific gene 6 and protein S genes and is involved in controlling cell survival and proliferation, spermatogenesis, immunoregulation and phagocytosis. The encoded protein has also been identified as a cell entry factor for Ebola and Marburg viruses.

Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 6D6F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR