Anti-ITK Mouse Monoclonal Antibody [clone: 5G12C4]

Supplier: Thermo Fisher Scientific
PIEAMA515410EA 0 EUR
PIEAMA515410
Anti-ITK Mouse Monoclonal Antibody [clone: 5G12C4]
Antibodies
MA5-15410 targets ITK in WB applications and shows reactivity with Human samples. The MA5-15410 immunogen is purified recombinant fragment of ITK (aa2-110) expressed in E. Coli. <br>. This gene encodes an intracellular tyrosine kinase expressed in T-cells. The protein contains both SH2 and SH3 domains which are often found in intracellular kinases. It is thought to play a role in T-cell proliferation and differentiation.

Type: Primary
Antigen: ITK
Clonality: Monoclonal
Clone: 5G12C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR