Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ALCAM Mouse Monoclonal Antibody [clone: 3F8B12]

Supplier: Thermo Scientific
MA5-15411 targets ALCAM in WB applications and shows reactivity with Human samples. The MA5-15411 immunogen is purified recombinant fragment of ALCAM (aa405-524) expressed in E. Coli. ALCAM(CD166): Activated leukocyte cell adhesion molecule.CD166 is a member of the Ig superfamily and is expressed on activated T-cells, B cells and other cells including thymic epithelial cells, fibroblasts, keratinocytes and neurons. CD6 has been identified as a receptor for CD166.

Type: Primary
Antigen: ALCAM
Clonality: Monoclonal
Clone: 3F8B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAMA515411 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ALCAM
Antigen name Activated Leukocyte Cell Adhesion Molecule
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody type Primary
Clone 3F8B12
Host Mouse
Isotype IgG2a
Western blot Yes
Antigen synonyms FLJ38514, CD166, ALCAM, MGC71733, MEMD
Gene ID 214
Format ascites
Immunogen Purified recombinant fragment of ALCAM (aa405-524) expressed in E. Coli.
Form Liquid
Storage buffer ascites, 0.03% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty