Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMAL1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: 5 ul of reticulocyte lysate translated hMOP3 protein, antigen standard for ARNTL (transient overexpression lysate). MOP3 (BMAL1), a general dimerization partner for a subset of the bHLH-PAS (basic-helix-loop-helix-PER-ARNT-SIM) superfamily of transcription regulators, is involved in circadian rhythm generation. MOP3 interacts with MOP4, CLOCK, hypoxia-inducible factor-1 alpha and -2 alpha. This interaction appears to be distinct from that of the more characterized general partner, ARNT (HIF-1Beta).

Type: Primary
Antigen: BMAL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
400 µL 400 µl PIEAPA116516 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol BMAL1
Antigen name Ayl hydrocarbon Receptor nuclear translocator-like
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoPrecipitation Yes
ChIP Yes
Antigen synonyms MOP3, TIC, JAP3, bHLHe5, BMAL1c, MGC47515, PASD3, BMAL1
Gene ID 406
Format whole serum
Immunogen Recombinant protein corresponding to residues 294-624 of human BMAL1.
Form Liquid
Storage buffer whole serum, 0.02% sodium azide
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping