Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PHD2 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody does not react with mouse samples. Suggested positive control: antigen standard for EGLN1 (transient overexpression lysate), human glioblastoma cells. HIF prolyl hydroxylase 2 is a prolyl hydroxylase that modifies HIF-alpha. Classic prolyl hydroxylases are found in the endoplasmic reticulum and modify collagen, whereas HIF is an intracellular protein and the HPH sites do not resemble those modifying collagen. HIF is a transcriptional complex that plays a critical role in oxygen homeostasis. HPH is an essential component of the pathway through which cells sense oxygen. In the presence of oxygen, HPHs convert specific prolyl residues in HIF-alpha to hydroxyproline, leading to HIF-alpha destruction. Low oxygen levels, sensed at the cellular level, cause the HIF conversion to be reduced so that HIF is stable and there is increased angiogenesis. HPH-2, specifically, catalyzes the posttranslational formation of 4-hydroxyproline in HIF alpha proteins. It hydroxylates HIF-1 alpha at Pro(402) and Pro(564), and HIF-2 alpha. It targets HIF through the hydroxylation for proteasomal degradation via the von Hippel-Lindau ubiquitylation complex.

Type: Primary
Antigen: PHD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat, Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116525 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PHD2
Antigen name HIF Prolyl Hydroxylase 2
Reactivity Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms HIFPH2, HIF-P4H 2, HIF Prolyl Hydroxylase 2, EGLN 1 EGLN1, Prolyl Hy, Chromosome 1 Open Reading Frame 12, P4H2, EGL 9 homolog of C. elegans 1, C1ORF12
Gene ID 308913, 54583
Concentration 1 mg/ml
Immunogen The epitope recognized by this antibody maps to a region between residues 1 and 50 of human PHD2/HIF Prolyl Hydroxylase 2 using the numbering given in entry NP_0713341 (GeneID 54583)
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris citrate, 0.09% sodium azide, pH 7-8
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping