Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Aromatase Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody is expected to react with equine (91%), bovine and rabbit (90%), porcine (83%), ovine, goat and canine (81%) based on sequence homology. Suggested positive control: antigen standard for CYP19A1 (transient overexpression lysate), human Fetal Temporal Lobe protein. Aromatase is a key enzyme in steroidogenesis and plays an important role in sexual differentiation, fertility, and carcinogenesis. Many environmental chemicals may influence aromatase activity and thereby disrupt endocrine function.

Type: Primary
Antigen: Aromatase
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit, Human, Primates

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116532 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol Aromatase
Antigen name Cytochrome P450 Family 19 Subfamily A Member 1
Reactivity Rabbit, Human, Primates
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoFluorescence Yes
Antigen synonyms Estrogen synthetase, CYP19A1, Cytochrome P450 19A1, CYPXIX, ARO1, CYP19, CYAR, P-450AROM
Gene ID 1588
Concentration 1.2 mg/ml
Immunogen Synthetic peptide made to a C-terminal portion of the human aromatase protein sequence (between residues 400-502).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris citrate, 0.1% sodium azide, pH 7-8
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty