Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FANCD2 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
In Western blot, this antibody should recognize a band at ~166 kDa (post-translationally modified form). Additional bands may be seen at lower molecular weights. In immunofluorescence, this has been tested in human MMC and IR-treated MEF cells.Suggested positive control: Hela whole cell extract. Fanconi anemia (FANC) is a human autosomal-recessive cancer susceptibility disorder characterized by congenital defects, progressive bone marrow failure, and cellular hypersensitivity to mitomycin C (MMC). The FANC subunit D2 protein is vital for cellular resistance to DNA cross-linking and the arrest of DNA synthesis after ionizing radiation. DNA damage activates the monoubiquitination of FANCD2, targeting nuclear foci containing the BRCA1 protein.

Type: Primary
Antigen: FANCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Primates, Mouse




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µL 50 µl PIEAPA116548 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol FANCD2
Antigen name Fanconi Anemia Complementation Group D2
Reactivity Primates, Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms FAD2, FANCONI ANEMIA COMPLEMENTATION GROUP D, FACD, FA4, FANCONI PANCYTOPENIA TYPE 4, FAD, FANCD
Gene ID 2177, 211651
Concentration 1 mg/ml
Immunogen Human FANCD2 fusion protein (N-terminal fragment).
Purification Affinity chromatography
Form Liquid
Storage buffer PBS, 0.05% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping