Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-RAD9 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
In Western blot, a band is seen at approximately 42 kDa, however multiple bands may be seen between ~40-60 kDa, depending on phosphorylation of the protein. Suggested positive control: antigen standard for RAD9A (transient overexpression lysate), Osteocarcinoma (U2OS) cell lysate. Human Rad 9 is a homologue of Schizosaccharomyces pombe Rad 9 protein and plays a critical role in cell cycle checkpoint control.

Type: Primary
Antigen: RAD9
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
50 µL 50 µl PIEAPA116553 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol RAD9
Antigen name RAD9 Checkpoint Clamp Component A
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms RAD 9 (S pombe) homolog, Cell cycle checkpoint control protein, RAD9 homolog, RAD9 homolog A, RAD9A
Gene ID 5883
Immunogen Fusion protein of full length human Rad 9 protein.
Purification Protein G
Form Liquid
Storage buffer tris glycine, 0.15M NaCl, 0.1% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty