Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PHD1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: antigen standard for EGLN2 (transient overexpression lysate), Hela whole cell lysate, HeLa and MEF extracts, MEF whole cell lysate. HIF prolyl hydroxylase 1 (HPH-3)is a prolyl hydroxylase that modifies HIF-alpha. Classic prolyl hydroxylases are found in the endoplasmic reticulum and modify collagen, whereas HIF is an intracellular protein and the HPH sites do not resemble those modifying collagen. HIF is a transcriptional complex that plays a critical role in oxygen homeostasis. HPH is an essential component of the pathway through which cells sense oxygen. In the presence of oxygen, HPHs convert specific prolyl residues in HIF-alpha to hydroxyproline, leading to HIF-alpha destruction. Low oxygen levels, sensed at the cellular level, cause the HIF conversion to be reduced so that HIF is stable and there is increased angiogenesis.

Type: Primary
Antigen: PHD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µl PIEAPA116569 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PHD1
Antigen name HIF Prolylhydroxylase 1
Reactivity Mouse, Rat, Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms estrogen-induced tag 6, prolyl hydroxylase domain-containing protein, HIF-P4H 1, P4H1, HIF prolyl hydroxylase 1, EGLN2, EGL nine (C.elegans) homolog 2
Gene ID 112398, 308457, 112406
Concentration 1 mg/ml
Immunogen A region between residue 350 and the C-terminus (residue 407) of human Prolyl Hydroxylase Domain-Containing Protein 1
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris citrate, 0.1% sodium azide, pH 7-8
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty