Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYR61 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody is predicted to react with rat and mouse based on 88% sequence homology. Suggested positive control: antigen standard for CYR61 (transient overexpression lysate), MDA-MB-231 breast cancer cell lysate. CYR61 is a cysteine-rich, heparin-binding, growth factor belonging to the CCN family of secreted regulatory proteins. It stimulates chondrogensis and angiogenesis, playing an important role in cartilage and bone formation, as well as tumor growth and vascularization.

Type: Primary
Antigen: CYR61
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116579 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol CYR61
Antigen name Insulin-like Growth Factor-binding Protein 10
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Neutralisation Yes
Antigen synonyms GIG1, Cysteine-rich angiogenic induce 61, CCN1, insulin-like growth factor-binding protein 10, IGFBP10
Gene ID 3491
Concentration 1 mg/ml
Immunogen A synthetic peptide made to the human CYR61 protein sequence (between residues 150-250).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.02% sodium azide
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty