Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PINK1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
This antibody has only been tested on transfected lysates. Endogenous protein detection is unknown. Mutations in the PTEN-induced kinase (PINK1) gene located within the PARK6 locus on chromosome 1p35-p36 have recently been identified in patients with recessive early-onset Parkinson disease. Data suggests that PINK1 may be the second most common causative gene next to parkin in parkinsonism with the recessive mode of inheritance.

Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG PIEAPA116625 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol PINK1
Antigen name PTEN-Induced Putative Kinase 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms PTEN induced putative kinase 1, BRPK, Serine/threonine kinase PINK1 mitochondrial, PINK 1, Phosphatase and Tensin Homolog, PINK-1, PARK6
Gene ID 65018
Concentration 1 mg/ml
Immunogen A synthetic peptide made residues 258-274 (YRKSKRGPKQLAPHPNI) of human PINK1.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.02% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty