Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-p14ARF Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
p14ARF tends to run slightly higher than the theoretical MW of 14 kDa. Suggested positive control: BT549 cells or HeLa whole cell extract, antigen standard for CDKN2A (transient overexpression lysate). The INK4a-ARF locus is comprised of two tumor suppressors, p16INK4a and p14ARF. These two proteins are encoded through differential splicing of alternative first exons. The p16INK4a (exon 1alpha) protein inhibits the cyclin D-dependent kinases (CDK) that control the phosphorylation of the Rb protein and cell proliferation. The p14ARF gene product complexes with the MDM2 protein within the nucleus, thus modulating the activity of the p53 protein. P14ARF is a potent tumor suppressor in the presence of wild-type p53, while mutant p53 substantially reduces growth inhibition by p14ARF.

Type: Primary
Antigen: p14ARF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116639 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol p14ARF
Antigen name Cyclin-dependent kinase inhibitor 2A
Reactivity Human, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms Cyclin dependent ki, CDKN2A, Cyclin dependent kinase inhibitor 2A, Cell cycle negative regulator beta, ARF, CDKN2, CDK4 inhibitor p16INK4, CDK4I, CMM2
Gene ID 1029, 12578
Concentration 1 mg/ml
Immunogen A synthetic peptide made to a portion of human p14ARF (between residues 50-150) .
Purification Antigen affinity chromatography
Form Liquid
Storage Buffer PBS, 0.05% sodium azide
Storage temperature -20°C or -80°C if preferred
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty