Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-p14ARF Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: BT549 or HeLa whole cell extracts, antigen standard for CDKN2A (transient overexpression lysate). The INK4a-ARF locus is comprised of two tumor suppressors, p16INK4a and p14ARF. These two proteins are encoded through differential splicing of alternative first exons. The p16INK4a (exon 1alpha) protein inhibits the cyclin D-dependent kinases (CDK) that control the phosphorylation of the Rb protein and cell proliferation. The p14ARF gene product complexes with the MDM2 protein within the nucleus, thus modulating the activity of the p53 protein. P14ARF is a potent tumor suppressor in the presence of wild-type p53, while mutant p53 substantially reduces growth inhibition by p14ARF.

Type: Primary
Antigen: p14ARF
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µl PIEAPA116662 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol p14ARF
Antigen name Cyclin-dependent kinase inhibitor 2A
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Antigen synonyms CDK4 inhibitor p16INK4, Cyclin dependent ki, CDKN2A, CMM2, Cell cycle negative regulator beta, CDK4I, CDKN2, ARF, Cyclin dependent kinase inhibitor 2A
Gene ID 1029
Concentration 1 mg/ml
Immunogen A synthetic peptide to human p14ARF (C-terminal sequence).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris citrate, 0.1% sodium azide, pH 7-8
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty