Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAP43 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: embryonic nervous tissue, brain, injured nerve. GAP-43 is expressed by developing and regenerating neurons, and to a lesser extent, reactive glial cells. It is used widely to specifically label injured neurons and to score neuronal regeneration.

Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chicken, Rat, Mouse

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116729 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol GAP43
Antigen name Growth associated protein 43
Reactivity Mouse, Rat, Human, Chicken
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms Nerve Growth Related Peptide, Growth Associated Protein 43, Neuromodulin
Gene ID 29423, 2596, 427955, 14432
Concentration 1 mg/ml
Immunogen Synthetic peptide, coupled to maleimide-activated KLH, corresponding to residues (C) K(216) E D P E A D Q E H A(226) of rat/mouse GAP-43.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.05% sodium azide
Storage temperature -20° C, Avoid Freeze/Thaw Cycles
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty