Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Laminin Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
The antibody functionally inhibits Laminin in mouse and rat. It binds to Laminin and inhibits most, if not all, of its cell adhesion and growth promotive properties. Immunostaining is enhanced by antigen retrieval with pepsin, especially paraffin tissue.Suggested positive control: basement membranes (blood vessels of any tissue, tumors, nerve). Laminin antibodies are widely used to label blood vessels and basement membranes.

Type: Primary
Antigen: Laminin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit, Rat, Ovine, Mouse, Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116730 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol Laminin
Antigen name Laminin
Reactivity Ovine, Rabbit, Mouse, Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms AA408497, Lama
Gene ID 16772, 284217, 101102692, 316758
Concentration 1 mg/ml
Immunogen Laminin isolated from mouse EHS tumor.
Purification IgG fraction
Form Liquid
Storage buffer PBS, 0.05% sodium azide
Storage temperature -20°C or -80°C if preferred
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping