Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Artemin Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: human dorsal root ganglia/CNS preparation. Artemin is a novel member of the GDNF family and has demonstrated that it is the ligand for the former orphan receptor GFRalpha3-RET. Artemin is a survival factor for sensory and sympathetic neurons in culture, and its expression pattern suggests that it also influences these neurons in vivo. Artemin can also activate the GFRalpha1-RET complex and supports the survival of dopaminergic midbrain neurons in culture.

Type: Primary
Antigen: Artemin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µg 100 µG PIEAPA116738 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol Artemin
Antigen name Artemin
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms EVN, NBN, ENOVIN
Gene ID 9048
Concentration 1 mg/ml
Immunogen Internal sequence of human Artemin.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.02% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty