Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SCP3 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Suggested positive control: mouse testis protein. The synaptonemal complex is a proteinaceous complex that apparently mediates synapses during the zygotene stage and then disintegrates. It is a complex structure that unites homologous chromosomes during the prophase stage of meiosis. It is the result of chromosome pairing interactions and not the cause of them.

Type: Primary
Antigen: SCP3
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Porcine, Chicken, Bovine, Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116766 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol SCP3
Antigen name Synaptonemal Complex Protein 3
Reactivity Porcine, Chicken, Mouse, Bovine, Human, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen synonyms Sycp3, COR1, RNASCP3, Synaptonemal complex protein 3
Gene ID 50511, 25561, 20962, 615896
Concentration 1 mg/ml
Immunogen A synthetic peptide made to the C-terminal region of the human SCP3 protein.
Purification Antigen affinity chromatography
Form Liquid
Storage buffer PBS, 0.09% sodium azide
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty