Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GAPDH Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: antigen standard for GAPDH (transient overexpression lysate). GAPDH is a 146 kDa tetramer composed of four 30-40 kDa subunits. Glyceraldehyde 3-Phosphate Dehydrogenase (GAPDH) is a metabolic enzyme responsible for catalyzing one step in the glycolytic pathway, the reversible oxidative phosphorylation of glyceraldehyde 3-phosphate. Because GAPDH as a protein expressed in large amounts and which is required at all times for an important house keeping functions, levels of GAPDH mRNA are often used as standards in studies of mRNA expression. Increasingly, scientists are making use of specific antibodies to GAPDH as loading controls for western blotting experiments. Apart from a role in glycolysis, GAPDH may have other roles such as in the activation of transcription. GAPDH is reported to bind to a variety of other proteins, including the amyloid precursor protein, mutations in which cause some forms of Alzheimer's disease, and the polyglutamine tracts of Huntingtin, the protein product aberrant forms of which are causative of Huntington's disease. Associations with actin and tubulin have also be reported. The protein may also have a role in the regulation of apoptosis, and interestingly migrates from the cytoplasm into the nucleus when cells become apoptotic.

Type: Primary
Antigen: GAPDH
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Primates, Rat, Mouse, Human

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116777 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol GAPDH
Antigen name Glyceraldehyde-3-phosphate dehydrogenase
Reactivity Rat, Mouse, Human, Primates
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Antigen synonyms GAPD, Glyceraldehyde-3-Phosphate Dehydrogenase, G3PD, G3PDH
Gene ID 14433, 24383, 2597
Concentration 0.2 mg/ml
Immunogen Residues between 150 and 200 of human Glyceraldehyde-3-Phosphate Dehydrogenase (GeneID 2597)
Purification Antigen affinity chromatography
Form Liquid
Storage buffer TBS, 0.1% BSA, 0.09% sodium azide
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty