Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ABCE1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific

Type: Primary
Antigen: ABCE1 (ATP-binding cassette, sub-family E (OABP), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Antibody size Pk VWR Catalog Number Quantity
100 µl 100 µl PIEAPA116797
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol ABCE1
Antigen name ATP Binding Cassette, Subfamily E, Member 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype none
Western Blot Yes
Antigen synonyms RNS4I, ABCE 1, ATP Binding Cassette Sub Family E Member 1, ABC 38, ABC38, RNase 4 inhibitor, RLI, Ribonuclease 4 Inhibitor, OABP, RNase L Inhibitor
Storage Buffer PBS with 0,05% Sodium azide
Storage temperature Cool
Shipping Temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty