Anti-LIPG Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
PIEAPA116799EA 0 EUR
PIEAPA116799
Anti-LIPG Rabbit Polyclonal Antibody
Antibodies
This antibody does not react with mouse samples. Endothelial lipase (EL) is a novel member of the lipoprotein lipase gene family. It is synthesized by endothelial cells and functions at the site of synthesis. EL most likely plays a physiological role in modulating lipoprotein metabolism. Overexpressed EL has been shown to reduce HDL cholesterol levels, whereas an EL deficiency appears to be effective in elevating HDL cholesterol levels in animals.

Type: Primary
Antigen: LIPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR