Anti-ChREBP Rabbit Polyclonal Antibody

Supplier: Thermo Fisher Scientific
PIEAPA116807EA 0 EUR
PIEAPA116807
Anti-ChREBP Rabbit Polyclonal Antibody
Antibodies
The target sequence has 93% sequence homology with mouse and 87% sequence homology with rat. ChREBP is a transcription factor involved in activating genes that encode enzymes of fatty acid biosynthesis in liver and adipose. This protein may have relevance to lipid metabolism, obesity, and type II diabetes.

Type: Primary
Antigen: ChREBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR