Anti-CD36 Rabbit Polyclonal Antibody (Biotin) [clone: ]

Supplier: Thermo Fisher Scientific
PIEAPA116815EA 0 EUR
PIEAPA116815
Anti-CD36 Rabbit Polyclonal Antibody (Biotin) [clone: ]
Antibodies
This antibody has 95% sequence homology with, rat, monkey, Golden hamster, and rabbit; 90% sequence homology with mouse and 75% with bovine. CD36 is a cell surface class B scavenger receptor. CD36 is preferentially found within lipid rafts, which facilitates its association with receptors, signalling and adapter molecules. CD36 binds to collagen, thrombospondin, anionic phospholipids and oxidized LDL. May function as a cell adhesion molecule. Directly mediates cyto adherence of Plasmodium falciparum parasitized erythrocytes. Binds long chain fatty acids and may function in the transport and/or as a regulator of fatty acid transport.

Type: Primary
Antigen: CD36
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR