Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NPC1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
By Western blot, PA1-16817 detects heterogeneously glycosylated NPC1 protein with prominent bands at ~170 and ~220 kDa. Suggested positive control: human fibroblast cell lysate. Niemann-Pick type C1 (NPC1) is a member of a family of genes encoding membrane-bound proteins containing putative sterol sensing domains. NPC1 protein regulates cholesterol transport from late endosomes-lysosomes to other intracellular compartments. NPC1 overexpression increases the rate of trafficking of low density lipoprotein cholesterol to the endoplasmic reticulum and the rate of delivery of endosomal cholesterol to the plasma membrane. NPC disease is an inherited neurovisceral lipid storage disorder of unesterified cholesterol accumulation in lysosomes. It is characterized by progressive neural and liver degeneration, resulting from inactivating mutations in NPC1, in most cases.

Type: Primary
Antigen: NPC1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Hamster, Human, Primates, Mouse, Porcine, Rat

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116817 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol NPC1
Antigen name Niemann-Pick disease type C1
Reactivity Rat, Porcine, Hamster, Mouse, Human, Primates
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Antigen synonyms NPC, FLJ98532
Gene ID 18145, 4864, 397591, 100689424
Concentration 1 mg/ml
Immunogen A synthetic peptide made to the C-terminal region of human NPC1.
Purification Affinity chromatography
Form Liquid
Storage buffer PBS, 0.1% sodium azide
Storage temperature Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty