Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LXR alpha Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
The target sequence has 92% sequence homology with rat and mouse, and 90% sequence homology with chicken. Suggested positive control: human liver lysate, human liver protein. Liver X receptor (LXR) is a protein involved in lipid metabolism. It plays a dual role in helping the body maintain a healthy balance between fat storage and the burning of fat.

Type: Primary
Antigen: LXR alpha
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Primates, Porcine

Log in to see your contract pricing and availability.
  Remember me on this device


Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116824 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page


Antigen symbol LXR alpha
Antigen name Nuclear Receptor subfamily 1 Group H Member 3
Reactivity Human, Porcine, Primates, Mouse
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms LXRb, NR1H2, Ubiquitously expressed nuclear receptor, NER, Liver X receptor alpha, Nuclear receptor NER, Liver X receptor beta, NERI, NR1H3, LXRa
Gene ID 10062, 22259
Concentration 1.0 mg/ml
Immunogen A synthetic peptide made to an internal portion of the human LXR protein sequence (between residues 50-150).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris glycine, 0.15M NaCl, 0.05% sodium azide
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty