Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POT1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
The target sequence has 95% sequence homology with monkey, 82% sequence homology with rat, and 72% sequence homology with mouse. Suggested positive control: HeLa nuclear extract, antigen standard for POT1 (transient overexpression lysate). Telomere maintenance is essential for protection against changes in telomere length, which has been implicated in ageing and certain cancers. POT1 (protection of telomeres 1), a telomere regulator, is a single-stranded telomeric DNA-binding protein that controls telomerase-mediated telomere elongation.

Type: Primary
Antigen: POT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Pk VWR Catalog Number Quantity
100 µL 100 µl PIEAPA116830 Blocked
Blocked This product has been blocked by your organisation. Please contact your purchasing department for more information.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen symbol POT1
Antigen name Protection of telomeres 1
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
Antigen synonyms POT 1, POT-1, Protection of telomeres 1
Gene ID 25913
Concentration 2.25 mg/ml
Immunogen A synthetic peptide made to an internal portion of the human POT1 protein sequence (between residues 200-300).
Purification Antigen affinity chromatography
Form Liquid
Storage buffer tris glycine, 0.15M NaCl, 0.05% sodium azide
Storage temperature 4° C, do not freeze
Shipping temperature Ice
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping